Learn More
Analytical Sales & Services Armor C18 Microbore Column. 120Â, 3um, 15cm x 1.0mm ID
Catalog No. 5000805757
Encompass
Armor C18 Microbore Column. 120Â, 3um, 15cm x 1.0mm ID
Catalog No. 50-008-05757 Supplier Analytical Sales & Services Supplier No. ADV15M1A203
Armor C18 Microbore Column. 120Â, 3um, 15cm x 1.0mm ID