Learn More
Analytical Sales & Services Armor C18 Microbore Column. 120Â, 5µm, 10cm x 0.3mm ID
Catalog No. 5000804576
Encompass
Armor C18 Microbore Column. 120Â, 5µm, 10cm x 0.3mm ID
Catalog No. 50-008-04576 Supplier Analytical Sales & Services Supplier No. ADV10M3A205
Add to Cart
Add to Cart
Armor C18 Microbore Column. 120Â, 5µm, 10cm x 0.3mm ID