Learn More
Analytical Sales & Services Armor C18 Microbore Column. 120Â, 5µm, 10cm x 0.5mm ID
Catalog No. 5000805886
Encompass
Armor C18 Microbore Column. 120Â, 5µm, 10cm x 0.5mm ID
Catalog No. 50-008-05886 Supplier Analytical Sales & Services Supplier No. ADV10M5A205
Armor C18 Microbore Column. 120Â, 5µm, 10cm x 0.5mm ID