Learn More
Analytical Sales & Services Armor C18 Microbore Column. 120Â, 5µm 10cm x 1.0 mm ID
Catalog No. 5000804575
Encompass
Armor C18 Microbore Column. 120Â, 5µm, 10cm x 1.0 mm ID
Catalog No. 50-008-04575 Supplier Analytical Sales & Services Supplier No. ADV10M1A205
Armor C18 Microbore Column. 120Â, 5µm, 10cm x 1.0 mm ID