Learn More
Analytical Sales & Services Armor C18 Microbore Column. 120Â, 5µm, 15cm x 1.0mm ID
Catalog No. 5000805961
Encompass
Armor C18 Microbore Column. 120Â, 5µm, 15cm x 1.0mm ID
Catalog No. 50-008-05961 Supplier Analytical Sales & Services Supplier No. ADV15M1A205
Armor C18 Microbore Column. 120Â, 5µm, 15cm x 1.0mm ID