Learn More
Analytical Sales & Services Armor C18 Microbore Column. 120Â, 5µm, 5cm x 0.3 ID
Catalog No. 5000805950
Encompass
Armor C18 Microbore Column. 120Â, 5µm, 5cm x 0.3 ID
Catalog No. 50-008-05950 Supplier Analytical Sales & Services Supplier No. ADV05M3A205
Armor C18 Microbore Column. 120Â, 5µm, 5cm x 0.3 ID