Learn More
Analytical Sales & Services Armor C18 Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID
Catalog No. 5000805864
Encompass
Armor C18 Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID
Catalog No. 50-008-05864 Supplier Analytical Sales & Services Supplier No. ADV05M5A205
Add to Cart
Add to Cart
Armor C18 Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID