Learn More
Analytical Sales & Services Armor C18 PEEK Guard Cartridges, 5µm, 20mm x 2.1mm ID
Catalog No. 5000805893
Encompass
Armor C18 PEEK Guard Cartridges, 5µm, 20mm x 2.1mm ID
Catalog No. 50-008-05893 Supplier Analytical Sales & Services Supplier No. ADVA185
Armor C18 PEEK Guard Cartridges, 5µm, 20mm x 2.1mm ID