Learn More
Analytical Sales & Services Armor C18 Prep Column. 120Â, 10µm, 25cm x 20mm ID (ADV712520)
Catalog No. 5000806667
Encompass
Armor C18 Prep Column. 120Â, 10µm, 25cm x 20mm ID
Catalog No. 50-008-06667 Supplier Analytical Sales & Services Supplier No. ADV712520
Armor C18 Prep Column. 120Â, 10µm, 25cm x 20mm ID