Learn More
Analytical Sales & Services Armor C18 Prep Column. 120Â, 5µm, 15cm x 20.0mm ID
Catalog No. 5000806666
Encompass
Armor C18 Prep Column. 120Â, 5µm, 15cm x 20mm ID
Catalog No. 50-008-06666 Supplier Analytical Sales & Services Supplier No. ADV721520
Armor C18 Prep Column. 120Â, 5µm, 15cm x 20mm ID