Learn More
Analytical Sales & Services Armor C18 Semi-Prep Column. 120Â, 10µm, 15cm x 10mm ID
Catalog No. 5000806161
Encompass
Armor C18 Semi-Prep Column. 120Â, 10µm, 15cm x 10mm ID
Catalog No. 50-008-06161 Supplier Analytical Sales & Services Supplier No. ADV711510
Armor C18 Semi-Prep Column. 120Â, 10µm, 15cm x 10mm ID