Learn More
Analytical Sales & Services Armor C18 Semi-Prep Column. 120Â, 5µm, 25cm x 10mm ID
Catalog No. 5000805722
Encompass
Armor C18 Semi-Prep Column. 120Â, 5µm, 25cm x 10mm ID
Catalog No. 50-008-05722 Supplier Analytical Sales & Services Supplier No. ADV722510
Armor C18 Semi-Prep Column. 120Â, 5µm, 25cm x 10mm ID