Learn More
Analytical Sales & Services Armor C8 Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID
Catalog No. 5000805451
Encompass
Armor C8 Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID
Catalog No. 50-008-05451 Supplier Analytical Sales & Services Supplier No. ADV5282
Armor C8 Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID