Learn More
Analytical Sales & Services Armor C8 Analytical Column. 120Â, 5µm, 10cm x 3.0mm ID
Catalog No. 5000805685
Encompass
Armor C8 Analytical Column. 120Â, 5µm, 10cm x 3.0mm ID
Catalog No. 50-008-05685 Supplier Analytical Sales & Services Supplier No. ADV5262
Armor C8 Analytical Column. 120Â, 5µm, 10cm x 3.0mm ID