Learn More
Analytical Sales & Services Armor C8 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID
Catalog No. 5000805603
Encompass
Armor C8 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID
Catalog No. 50-008-05603 Supplier Analytical Sales & Services Supplier No. ADV5216
Armor C8 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID