Learn More
Analytical Sales & Services Armor C8 Analytical Column. 120Â, 5µm, 15cm x 3.0mm ID
Catalog No. 5000806658
Encompass
Armor C8 Analytical Column. 120Â, 5µm, 15cm x 3.0mm ID
Catalog No. 50-008-06658 Supplier Analytical Sales & Services Supplier No. ADV5263
Add to Cart
Add to Cart
Armor C8 Analytical Column. 120Â, 5µm, 15cm x 3.0mm ID