Learn More
Analytical Sales & Services Armor C8 Analytical Column. 120Â, 5µm, 25cm x 3.0mm ID
Catalog No. 5000805996
Encompass
Armor C8 Analytical Column. 120Â, 5µm, 25cm x 3.0mm ID
Catalog No. 50-008-05996 Supplier Analytical Sales & Services Supplier No. ADV5264
Armor C8 Analytical Column. 120Â, 5µm, 25cm x 3.0mm ID