Learn More
Analytical Sales & Services Armor C8 Analytical Column. 120Â, 5µm, 3cm x 4.6mm ID
Catalog No. 5000806711
Encompass
Armor C8 Analytical Column. 120Â, 5µm, 3cm x 4.6mm ID
Catalog No. 50-008-06711 Supplier Analytical Sales & Services Supplier No. ADV5214
Armor C8 Analytical Column. 120Â, 5µm, 3cm x 4.6mm ID