Learn More
Analytical Sales & Services Armor C8 Analytical Column. 120Â, 5µm, 5cm x 2.1mm ID
Catalog No. 5000806155
Encompass
Armor C8 Analytical Column. 120Â, 5µm, 5cm x 2.1mm ID
Catalog No. 50-008-06155 Supplier Analytical Sales & Services Supplier No. ADV5281
Armor C8 Analytical Column. 120Â, 5µm, 5cm x 2.1mm ID