Learn More
Analytical Sales & Services Armor C8 Microbore Column. 120Â, 5µm, 10cm x 0.5mm ID
Catalog No. 5000805823
Encompass
Armor C8 Microbore Column. 120Â, 5µm, 10cm x 0.5mm ID
Catalog No. 50-008-05823 Supplier Analytical Sales & Services Supplier No. ADV10M5B205
Armor C8 Microbore Column. 120Â, 5µm, 10cm x 0.5mm ID