Learn More
Analytical Sales & Services Armor C8 Microbore Column. 120Â, 5µm, 10cm x 1.0mm ID
Catalog No. 5000805824
Encompass
Armor C8 Microbore Column. 120Â, 5µm, 10cm x 1.0mm ID
Catalog No. 50-008-05824 Supplier Analytical Sales & Services Supplier No. ADV10M1B205
Armor C8 Microbore Column. 120Â, 5µm, 10cm x 1.0mm ID