Learn More
Analytical Sales & Services Armor C8 Microbore Column. 120Â, 5µm, 15cm x 1.0mm ID
Catalog No. 5000806146
Encompass
Armor C8 Microbore Column. 120Â, 5µm, 15cm x 1.0mm ID
Catalog No. 50-008-06146 Supplier Analytical Sales & Services Supplier No. ADV15M1B205
Add to Cart
Add to Cart
Armor C8 Microbore Column. 120Â, 5µm, 15cm x 1.0mm ID