Learn More
Analytical Sales & Services Armor C8 Microbore Column. 120Â, 5µm, 3cm x 0.3mm ID
Catalog No. 5000805942
Encompass
Armor C8 Microbore Column. 120Â, 5µm, 3cm x 0.3mm ID
Catalog No. 50-008-05942 Supplier Analytical Sales & Services Supplier No. ADV03M3B205
Armor C8 Microbore Column. 120Â, 5µm, 3cm x 0.3mm ID