Learn More
Analytical Sales & Services Armor C8 Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID
Catalog No. 5000805925
Encompass
Armor C8 Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID
Catalog No. 50-008-05925 Supplier Analytical Sales & Services Supplier No. ADV05M5B205
Armor C8 Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID