Learn More
Analytical Sales & Services Armor C8 Microbore Column. 120Â, 5um, 25cm x 1.0mm ID
Catalog No. 5000805427
Encompass
Armor C8 Microbore Column. 120Â, 5um, 25cm x 1.0mm ID
Catalog No. 50-008-05427 Supplier Analytical Sales & Services Supplier No. ADV25M1B205
Armor C8 Microbore Column. 120Â, 5um, 25cm x 1.0mm ID