Learn More
Analytical Sales & Services Armor C8 Prep Column. 120Â, 5µm, 10cm x 20.0mm ID
Catalog No. 5000805635
Encompass
Armor C8 Prep Column. 120Â, 5µm, 10cm x 20mm ID
Catalog No. 50-008-05635 Supplier Analytical Sales & Services Supplier No. ADV731020
Armor C8 Prep Column. 120Â, 5µm, 10cm x 20mm ID