Learn More
Analytical Sales & Services Armor C8 Semi-Prep Column. 120Â, 5µm, 10cm x 10.0mm ID
Catalog No. 5000805463
Encompass
Armor C8 Semi-Prep Column. 120Â, 5µm, 10cm x 10mm ID
Catalog No. 50-008-05463 Supplier Analytical Sales & Services Supplier No. ADV731010
Armor C8 Semi-Prep Column. 120Â, 5µm, 10cm x 10mm ID