Learn More
Analytical Sales & Services Armor C8 Semi-Prep Column. 120Â, 5µm, 15cm x 10.0mm ID
Catalog No. 5000806641
Encompass
Armor C8 Semi-Prep Column. 120Â, 5µm, 15cm x 10mm ID
Catalog No. 50-008-06641 Supplier Analytical Sales & Services Supplier No. ADV731510
Armor C8 Semi-Prep Column. 120Â, 5µm, 15cm x 10mm ID