Learn More
MSC Armor Guys Kyorene Pro, A3 Cut, MicroFoam Nitrile, 6 Dozen/Case
Catalog No. 5000770286
Encompass
The glove features an 18g Gray Kyorene Pro Graphene A3 Liner and a Black HCT MicroFoam Nitrile Palm Coating, offering ANSI Cut Level A3 protection. It complies with FDA Food Handling Requirements (21 CFR 177.2600) and is available in various sizes.
  • Silicone free
  • Touchscreen compatible
  • Reinforced thumb crotch for extended wear
  • No stainless steel or fiberglass
  • Ergonomically shaped for superior fit
  • Bacteriostatic kills harmful bacteria
  • Thermal regulating to keep the hands comfortable
  • Odor neutralizing to keep the gloves smelling fresh
  • Cut resistance from level A up to F (EN 388) and A1 up to A9 (ANSI)
  • Abrasion resistance from level 1 to 4 (EN 388) and level 1 up to 6 (ANSI)
  • UV protection against UV-A and UV-B rays
Catalog No. 50-007-70286 Supplier MSC Supplier No. 13899851
The glove features an 18g Gray Kyorene Pro Graphene A3 Liner and a Black HCT MicroFoam Nitrile Palm Coating, offering ANSI Cut Level A3 protection. It complies with FDA Food Handling Requirements (21 CFR 177.2600) and is available in various sizes.
  • Silicone free
  • Touchscreen compatible
  • Reinforced thumb crotch for extended wear
  • No stainless steel or fiberglass
  • Ergonomically shaped for superior fit
  • Bacteriostatic kills harmful bacteria
  • Thermal regulating to keep the hands comfortable
  • Odor neutralizing to keep the gloves smelling fresh
  • Cut resistance from level A up to F (EN 388) and A1 up to A9 (ANSI)
  • Abrasion resistance from level 1 to 4 (EN 388) and level 1 up to 6 (ANSI)
  • UV protection against UV-A and UV-B rays