Learn More
Analytical Sales & Services Armor Semi-Prep SS C18 Guard Cartridge. 10µm, 3cm x 10mm ID
Catalog No. 5000805891
Encompass
Advantage™ Armor Semi-Prep Stainless Steel C18 Guard Cartridge. 10µm, 3cm x 10mm ID
Catalog No. 50-008-05891 Supplier Analytical Sales & Services Supplier No. ADV0310171
Advantage™ Armor Semi-Prep Stainless Steel C18 Guard Cartridge. 10µm, 3cm x 10mm ID