Learn More
Asuragen Inc Armored RNA Quant RNase P
Supplier:  Asuragen Inc 52031
Encompass
It is an in vitro transcribed RNA encoding the RNase P gene that is encapsulated in phage coat protein particles. This sequence aligns with the CDC Diagnostic Panel recommendation.
Catalog No. NC1805784