Learn More
Asuragen Inc Armored RNA Quant SARS-CoV-2 Control
Catalog No. NC1805783
Encompass
Armored RNA Quant controls can be added directly to human matrices as an exogenous internal control for extraction with most nucleic acid isolation methods and detection in molecular assays.
Catalog No. NC1805783 Supplier Asuragen Inc Supplier No. 52030
Armored RNA Quant controls can be added directly to human matrices as an exogenous internal control for extraction with most nucleic acid isolation methods and detection in molecular assays.