Learn More
Braintree Scientific Armour Touch Metal Mesh? Gloves,Medium, Size 9
SDP
Catalog No. NC1611468
Encompass
Armour Touch Metal Mesh Glove - Protects hand from teeth and claws when handling small animals. YES! You have seen this material used for anti-shark suits. Braintree Scientific's metal mesh gloves provide security and a sure grip. This is the original and only metal mesh glove for rodent handling. Experience shows animals are easier to control when hWheatons are relaxed and confident. Much better than leather since metal mesh is more flexible, is readily cleaned, won't stain or corrode and doesn't carry odors. Gloves may be worn over surgical gloves. Adjustable buckle wrist band prevents slipping. Convert right to left by turning inside out. Order by size. * Price is per glove **Sold for research purposes only and not for human use**
This item is not returnable. View return policy
Catalog No. NC1611468 Supplier Braintree Scientific Supplier No. MMG100M
Add to Cart
Add to Cart
Armour Touch Metal Mesh Glove - Protects hand from teeth and claws when handling small animals. YES! You have seen this material used for anti-shark suits. Braintree Scientific's metal mesh gloves provide security and a sure grip. This is the original and only metal mesh glove for rodent handling. Experience shows animals are easier to control when hWheatons are relaxed and confident. Much better than leather since metal mesh is more flexible, is readily cleaned, won't stain or corrode and doesn't carry odors. Gloves may be worn over surgical gloves. Adjustable buckle wrist band prevents slipping. Convert right to left by turning inside out. Order by size. * Price is per glove **Sold for research purposes only and not for human use**