Learn More
Medchemexpress LLC Arn 077 enantiomer | 1439366-88-7 | 99.7% | 1 MG
SDP
Catalog No. 5000231162
Encompass_Preferred
Arn 077 enantiomer is the less active enantiomer of ARN 077 and functions as an inhibitor of N-acylethanolamine acid amidase (NAAA); reported IC50 for rat NAAA is 3.53 μM. Supplied as a solid for research applications.
  • Less active enantiomer with reported IC50 = 3.53 μM for rat NAAA.
  • Chemical formula C16H21NO4; molecular weight 291.34 g/mol.
  • White to off-white solid; powder storage at -20°C for long-term stability.
  • High DMSO solubility (100 mg/mL); vehicle formulations reported for in vivo use at ≥ 2.5 mg/mL.
  • For research use only; handle according to appropriate safety procedures.
Catalog No. 50-002-31162 Supplier Medchemexpress LLC Supplier No. HY120813A1MG
Arn 077 enantiomer is the less active enantiomer of ARN 077 and functions as an inhibitor of N-acylethanolamine acid amidase (NAAA); reported IC50 for rat NAAA is 3.53 μM. Supplied as a solid for research applications.
  • Less active enantiomer with reported IC50 = 3.53 μM for rat NAAA.
  • Chemical formula C16H21NO4; molecular weight 291.34 g/mol.
  • White to off-white solid; powder storage at -20°C for long-term stability.
  • High DMSO solubility (100 mg/mL); vehicle formulations reported for in vivo use at ≥ 2.5 mg/mL.
  • For research use only; handle according to appropriate safety procedures.