Learn More
Medchemexpress LLC Arn 077 (URB913) | 1373625-34-3 | 98.8% | 25 MG
SDP
Catalog No. 5000231653
Encompass_Preferred
ARN 077 (URB913) is a potent, selective N-acylethanolamine acid amidase (NAAA) inhibitor supplied as a solid for research use; it is used in vitro and in vivo to increase palmitoyl ethanolamine (PEA) levels and to evaluate antinociceptive effects.
  • Selective NAAA inhibitor for neuroscience research.
  • High purity (~98.8%), white to off-white solid.
  • Soluble in DMSO; multiple in vivo formulation protocols reported.
  • Powder storage: -20°C up to 3 years, 4°C up to 2 years; in solvent: -80°C for 6 months.
  • Available in small research pack sizes including 25 MG.
Catalog No. 50-002-31653 Supplier Medchemexpress LLC Supplier No. HY12081325MG
ARN 077 (URB913) is a potent, selective N-acylethanolamine acid amidase (NAAA) inhibitor supplied as a solid for research use; it is used in vitro and in vivo to increase palmitoyl ethanolamine (PEA) levels and to evaluate antinociceptive effects.
  • Selective NAAA inhibitor for neuroscience research.
  • High purity (~98.8%), white to off-white solid.
  • Soluble in DMSO; multiple in vivo formulation protocols reported.
  • Powder storage: -20°C up to 3 years, 4°C up to 2 years; in solvent: -80°C for 6 months.
  • Available in small research pack sizes including 25 MG.