Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arn 077 (URB913) | 1373625-34-3 | 98.8% | 50 MG
SDP

Catalog No. 5000232271
Encompass_Preferred

ARN 077 (URB913) is a potent and selective N-acylethanolamine acid amidase (NAAA) inhibitor used as a research compound to modulate palmitoylethanolamide (PEA) levels in vitro and in vivo.

  • Potent NAAA inhibition with low nanomolar activity.
  • High purity (98.8%).
  • Molecular formula C16H21NO4, molecular weight 291.34.
  • Soluble in DMSO; in vitro solubility up to 250 mg/mL.
  • Stable as a powder at -20°C for long-term storage.
  • Supplied as a 50 mg solid for laboratory use.

Catalog No. 50-002-32271 Supplier Medchemexpress LLC Supplier No. HY12081350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ARN 077 (URB913) is a potent and selective N-acylethanolamine acid amidase (NAAA) inhibitor used as a research compound to modulate palmitoylethanolamide (PEA) levels in vitro and in vivo.

  • Potent NAAA inhibition with low nanomolar activity.
  • High purity (98.8%).
  • Molecular formula C16H21NO4, molecular weight 291.34.
  • Soluble in DMSO; in vitro solubility up to 250 mg/mL.
  • Stable as a powder at -20°C for long-term storage.
  • Supplied as a 50 mg solid for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.