Learn More
Medchemexpress LLC ARN-21934 | 2230854-93-8 | 98.1% | 360.46 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13746650MG
Encompass_Preferred
ARN-21934 is a potent, highly selective, and blood-brain barrier (BBB) penetrant inhibitor specifically targeting human topoisomerase IIα over β. It inhibits DNA relaxation with an IC50 of 2 μM, significantly more potent than the anticancer agent Etoposide (IC50=120 μM). The compound demonstrates a favorable in vivo pharmacokinetic profile and shows antiproliferative activity against various human cancer cell lines, making it a promising lead compound for anticancer research.
  • Potent and highly selective topoisomerase IIα inhibitor
  • Blood-brain barrier penetrant
  • Inhibits DNA relaxation with IC50 of 2 μM
  • Exhibits antiproliferative activity against human cancer cell lines
  • Favorable in vivo pharmacokinetic profile
  • Appears as a solid, light yellow to green yellow
  • Soluble in DMSO (8.33 mg/mL at 80°C with ultrasonic)
Catalog No. 50-003-48843