Learn More
Medchemexpress LLC ARN-21934 | 2230854-93-8 | 98.1% | 360.46 g·mol⁻1 | C21H24N6 | 5 MG
SDP
Catalog No. 5000222151
Encompass_Preferred
ARN-21934 is a selective small-molecule inhibitor of human topoisomerase IIα with demonstrated blood-brain barrier penetration. It inhibits DNA relaxation activity and is supplied for research use in solid form and as DMSO solutions; molecular data and CAS are provided for compound characterization.
  • Selective inhibition of human topoisomerase IIα
  • Demonstrated blood-brain barrier penetration
  • Supplied as solid and as 10 mM DMSO solutions
  • High purity suitable for biochemical and cellular studies
  • Molecular formula and weight documented for compound characterization
  • Available in multiple pack sizes to support varied study scales
Catalog No. 50-002-22151 Supplier Medchemexpress LLC Supplier No. HY1374665MG
ARN-21934 is a selective small-molecule inhibitor of human topoisomerase IIα with demonstrated blood-brain barrier penetration. It inhibits DNA relaxation activity and is supplied for research use in solid form and as DMSO solutions; molecular data and CAS are provided for compound characterization.
  • Selective inhibition of human topoisomerase IIα
  • Demonstrated blood-brain barrier penetration
  • Supplied as solid and as 10 mM DMSO solutions
  • High purity suitable for biochemical and cellular studies
  • Molecular formula and weight documented for compound characterization
  • Available in multiple pack sizes to support varied study scales