Learn More
Selleck Chemical LLC ARN-3236
SDP
Supplier:  Selleck Chemical LLC S85435MG
Encompass_Preferred
ARN-3236 is a potent orally available and selective inhibitor of salt-inducible kinase 2 (SIK2) with IC50 of 1 nM 21 63 nM and 6 63 nM for SIK2 SIK1 and SIK3 respectively ARN-3236 induces apoptosis in cancer cells
Catalog No. 50-217-3371