Learn More
Apexbio Technology LLC ARN-509, 10mg. Cas: 956104-40-8 MFCD: MFCD22380626. Androgen receptor inhibitor
SDP
Catalog No. 501905128
Encompass_Preferred
ARN-509, a synthetic biaryl thiohydantoin compound, is a competitive androgen receptor (AR) inhibitor and fully antagonistic to AR overexpression. The IC50 of ARN-509 is 16 nmol/L [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5128 Supplier Apexbio Technology LLC Supplier No. A836410
ARN-509, a synthetic biaryl thiohydantoin compound, is a competitive androgen receptor (AR) inhibitor and fully antagonistic to AR overexpression. The IC50 of ARN-509 is 16 nmol/L [1]. Other sizes are available. Please inqury us for quote.