Learn More
Medchemexpress LLC ARN19702 | 1971937-18-4 | 99.8% | C21H22FN3O3S2 | 50 MG
SDP
Catalog No. 5000355012
Encompass_Preferred
ARN19702 is a selective, orally active, reversible, and brain-penetrant N-acylethanolamine acid amidase (NAAA) inhibitor with an IC50 of 230 nM for human NAAA. It demonstrates pain relief effects and protective effects against multiple sclerosis.
  • Selective, orally active, reversible, and brain-penetrant N-acylethanolamine acid amidase (NAAA) inhibitor
  • IC50 of 230 nM for human NAAA
  • Demonstrates pain relief effects
  • Reduced nociception in paclitaxel-induced neuropathy models
  • Attenuated nocifensive responses and hypersensitivity in various pain models
  • Provided protective effects against multiple sclerosis
  • White to off-white solid appearance
  • Molecular weight of 447.55
Catalog No. 50-003-55012 Supplier Medchemexpress LLC Supplier No. HY14533950MG
ARN19702 is a selective, orally active, reversible, and brain-penetrant N-acylethanolamine acid amidase (NAAA) inhibitor with an IC50 of 230 nM for human NAAA. It demonstrates pain relief effects and protective effects against multiple sclerosis.
  • Selective, orally active, reversible, and brain-penetrant N-acylethanolamine acid amidase (NAAA) inhibitor
  • IC50 of 230 nM for human NAAA
  • Demonstrates pain relief effects
  • Reduced nociception in paclitaxel-induced neuropathy models
  • Attenuated nocifensive responses and hypersensitivity in various pain models
  • Provided protective effects against multiple sclerosis
  • White to off-white solid appearance
  • Molecular weight of 447.55