Learn More
Medchemexpress LLC ARN19874 | 2165322-94-9 | 98.0% | 422.21 g/mol | C17H16BrF4NO2 | 25 MG
SDP
Catalog No. 5000227968
Encompass_Preferred
USP25/28 inhibitor AZ1 is a selective, orally active, noncompetitive dual inhibitor of deubiquitinases USP25 and USP28, with reported IC50 values of approximately 0.6-0.7 μM. It is supplied for research use as a solid or as a 10 mM solution in DMSO, and is typically provided at 98% purity.
  • Selective dual inhibitor of USP25 and USP28
  • Noncompetitive mechanism with IC50 ~0.6-0.7 μM
  • Suitable for in vitro and in vivo preclinical studies
  • Available as solid and 10 mM solution in DMSO
  • High purity (98%) for research applications
Catalog No. 50-002-27968 Supplier Medchemexpress LLC Supplier No. HY11737025MG
USP25/28 inhibitor AZ1 is a selective, orally active, noncompetitive dual inhibitor of deubiquitinases USP25 and USP28, with reported IC50 values of approximately 0.6-0.7 μM. It is supplied for research use as a solid or as a 10 mM solution in DMSO, and is typically provided at 98% purity.
  • Selective dual inhibitor of USP25 and USP28
  • Noncompetitive mechanism with IC50 ~0.6-0.7 μM
  • Suitable for in vitro and in vivo preclinical studies
  • Available as solid and 10 mM solution in DMSO
  • High purity (98%) for research applications