Learn More
TARGETMOL CHEMICALS INC ARN25068 5MG
SDP
Catalog No. 502579525
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ARN25068 is a potent inhibitor of GSK-3(beta) FYN and DYRK1A protein kinases exerting its activity in the sub-micromolar range. This compound effectively addresses tau hyperphosphorylation [1]. purity: 99%
Catalog No. 50-257-9525 Supplier TARGETMOL CHEMICALS INC Supplier No. T613605MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ARN25068 is a potent inhibitor of GSK-3(beta) FYN and DYRK1A protein kinases exerting its activity in the sub-micromolar range. This compound effectively addresses tau hyperphosphorylation [1]. purity: 99%