Learn More
Cayman Chemical ArnbIae radIx ShIkonIn 100mg
SDP
Catalog No. 502971876
Encompass_Preferred
A naturally occurring naphthoquinine; increases glucose uptake by adipocytes and inhibits PTEN activity; inhibits glycolysis in cancer cells by inhibiting tumor-specific pyruvate kinase M2; inhibits leukocyte migration, downregulates chemokine receptor expression, and inhibits HIV-1 replication; reduces joint swelling and cartilage destruction in a mouse model of collagen-induced arthritis
Catalog No. 50-297-1876 Supplier Cayman Chemical Supplier No. 14751100 MG
A naturally occurring naphthoquinine; increases glucose uptake by adipocytes and inhibits PTEN activity; inhibits glycolysis in cancer cells by inhibiting tumor-specific pyruvate kinase M2; inhibits leukocyte migration, downregulates chemokine receptor expression, and inhibits HIV-1 replication; reduces joint swelling and cartilage destruction in a mouse model of collagen-induced arthritis