Learn More
Cayman Chemical Shikonin, Purity: ≥98%, CAS: 517-89-5, 10 mg
SDP
Catalog No. 502970095
Encompass_Preferred
A naturally occurring naphthoquinine; increases glucose uptake by adipocytes and inhibits PTEN activity; inhibits glycolysis in cancer cells by inhibiting tumor-specific pyruvate kinase M2; inhibits leukocyte migration, downregulates chemokine receptor expression, and inhibits HIV-1 replication; reduces joint swelling and cartilage destruction in a mouse model of collagen-induced arthritis
Catalog No. 50-297-0095 Supplier Cayman Chemical Supplier No. 1475110 MG
A naturally occurring naphthoquinine; increases glucose uptake by adipocytes and inhibits PTEN activity; inhibits glycolysis in cancer cells by inhibiting tumor-specific pyruvate kinase M2; inhibits leukocyte migration, downregulates chemokine receptor expression, and inhibits HIV-1 replication; reduces joint swelling and cartilage destruction in a mouse model of collagen-induced arthritis