Learn More
Cayman Chemical ArncIamycIn A 2.5mg
SDP
Catalog No. 502991069
Encompass_Preferred
An antibiotic; active against M. bovis and B. subtilis (MICs = 30 and 7.5 µM, respectively)
Catalog No. 50-299-1069 Supplier Cayman Chemical Supplier No. 316592.5 MG
An antibiotic; active against M. bovis and B. subtilis (MICs = 30 and 7.5 µM, respectively)