Learn More
Cayman Chemical ArnIcolIde D 10mg
SDP
Supplier:  Cayman Chemical 3619310 MG
Encompass_Preferred
A sesquiterpene lactone with anticancer activity; inhibits the proliferation of MDA-MB-231, MDA-MB-468, and MCF-7 human breast cancer cells (IC50s = 5.211, 3.405, and 9.083 µM, respectively, after 48 hours); inhibits migration of MDA-MB-231 cells at 2.5, 5, and 10 µM; reduces tumor volume in an MDA-MB-231 orthotopic mouse xenograft model at 25 and 50 mg/kg
Catalog No. 50-291-2272