Learn More
TARGETMOL CHEMICALS INC Arnicolide D 1MG
SDP
Catalog No. 502877042
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Arnicolide D is a sesquiterpene lactone. Arnicolide D isolates from Centipeda minima. Arnicolide D modulates the cell cycle, activates the caspase signaling pathway and inhibits the PI3K/AKT/mTOR and STAT3 signaling pathways. Arnicolide D inhibits Nasopharyngeal carcinoma (NPC) cell viability in a concentration- and time-dependent manner. Arnicolide D exerts strong cytotoxic activity on the human colon carcinoma HT-29 cell line. Purity 96.66%
Catalog No. 50-287-7042 Supplier TARGETMOL CHEMICALS INC Supplier No. TN14051MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Arnicolide D is a sesquiterpene lactone. Arnicolide D isolates from Centipeda minima. Arnicolide D modulates the cell cycle, activates the caspase signaling pathway and inhibits the PI3K/AKT/mTOR and STAT3 signaling pathways. Arnicolide D inhibits Nasopharyngeal carcinoma (NPC) cell viability in a concentration- and time-dependent manner. Arnicolide D exerts strong cytotoxic activity on the human colon carcinoma HT-29 cell line. Purity 96.66%