Learn More
Medchemexpress LLC Arnicolide D | 34532-68-8 | 10 MG
SDP
Catalog No. 5000421017
Encompass_Preferred
Arnicolide D is a sesquiterpene lactone isolated from Centipeda minima. It demonstrates cytotoxicity against tumor cells, inducing cell cycle arrest, apoptosis, and oncosis. Arnicolide D exhibits anti-tumor activity.
  • Antitumor agent: Is cytotoxic to tumor cells and has anti-tumor activity.
  • Induces cell cycle arrest and apoptosis in nasopharyngeal carcinoma cells by activating the caspase signaling pathway and inhibiting the PI3K/AKT/mTOR and STAT3 signaling pathways.
  • Induces endoplasmic reticulum stress-mediated and ROS-dependent oncosis in hepatocellular carcinoma cells via ATF4 and CHOP.
  • Demonstrates anti-melanoma effects and inhibits the NF-κB pathway in an in vivo B16F10 allograft mouse model.
  • High purity of 99.69%.
  • Solid, white to off-white appearance.
Catalog No. 50-004-21017 Supplier Medchemexpress LLC Supplier No. HYN684310MG
Arnicolide D is a sesquiterpene lactone isolated from Centipeda minima. It demonstrates cytotoxicity against tumor cells, inducing cell cycle arrest, apoptosis, and oncosis. Arnicolide D exhibits anti-tumor activity.
  • Antitumor agent: Is cytotoxic to tumor cells and has anti-tumor activity.
  • Induces cell cycle arrest and apoptosis in nasopharyngeal carcinoma cells by activating the caspase signaling pathway and inhibiting the PI3K/AKT/mTOR and STAT3 signaling pathways.
  • Induces endoplasmic reticulum stress-mediated and ROS-dependent oncosis in hepatocellular carcinoma cells via ATF4 and CHOP.
  • Demonstrates anti-melanoma effects and inhibits the NF-κB pathway in an in vivo B16F10 allograft mouse model.
  • High purity of 99.69%.
  • Solid, white to off-white appearance.