Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ArnicolideC | 34532-67-7 | 5 MG
SDP

Catalog No. 5000420154
Encompass_Preferred

ArnicolideC is a sesquiterpene lactone isolated from *Centipeda minima*. It demonstrates a cytotoxic effect on nasopharyngeal carcinoma (NPC) cells, significantly inhibiting cell growth in a dose- and time-dependent manner and exhibiting inhibitory effects on NPC proliferation.

  • Molecular weight: 334.41
  • Formula: C₁₉H₂₆O₅
  • Appearance: Solid, white to off-white
  • Structure classification: Terpenoids, sesquiterpenes
  • Purity: 99.81%
  • Isolated from *Centipeda minima*
  • Cytotoxic effect on nasopharyngeal carcinoma (NPC) cells
  • Inhibits cell growth in a dose- and time-dependent manner
  • Inhibits NPC proliferation

Catalog No. 50-004-20154 Supplier Medchemexpress LLC Supplier No. HYN68425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ArnicolideC is a sesquiterpene lactone isolated from *Centipeda minima*. It demonstrates a cytotoxic effect on nasopharyngeal carcinoma (NPC) cells, significantly inhibiting cell growth in a dose- and time-dependent manner and exhibiting inhibitory effects on NPC proliferation.

  • Molecular weight: 334.41
  • Formula: C₁₉H₂₆O₅
  • Appearance: Solid, white to off-white
  • Structure classification: Terpenoids, sesquiterpenes
  • Purity: 99.81%
  • Isolated from *Centipeda minima*
  • Cytotoxic effect on nasopharyngeal carcinoma (NPC) cells
  • Inhibits cell growth in a dose- and time-dependent manner
  • Inhibits NPC proliferation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.